siRNA Details
Gene Symbol: Fstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024369.2 | NP_077345.1 | 3 | 167 | ||
GenBank mRNA | AY325203.1 | - | 266 | |||
BC087014.1 | - | 3 | 167 | 7189359 | ||
U06864.1 | - | 3 | 142 |

More Information
Other Species Targeted

Gene Ontology Categories:
