siRNA Details
Gene Symbol: Grid2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024379.1 | NP_077355.1 | 1, 2 | 410 | ||
GenBank mRNA | U08256.1 | - | 1, 2 | 410 | ||
Z17239.1 | - | 1, 2 | 120 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008167, BC139823, AK036192, D13266, AK045348, and others.
Gene Ontology Categories:
