siRNA Details
Gene Symbol: Gk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004077.1 | NP_001004077.1 | 1 | 446 | ||
GenBank mRNA | BC079449.1 | - | 1 | 446 | 7115796 |
Gene Symbol: Gk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024381.2 | NP_077357.2 | 6 | 528 | ||
XM_006256987.2 | XP_006257049.1 | 6 | 562 | |||
XM_006256988.2 | XP_006257050.1 | 6 | 562 | |||
XM_008773274.1 | XP_008771496.1 | 7 | 627 | |||
XM_008773275.1 | XP_008771497.1 | 7 | 627 | |||
XM_008773276.1 | XP_008771498.1 | 7 | 627 | |||
XM_008773278.1 | XP_008771500.1 | 7 | 626 | |||
GenBank mRNA | D16102.1 | - | 6 | 531 |

More Information
Other Species Targeted

Gene Ontology Categories:
