siRNA Details
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024394.2 | NP_077370.2 | 5 | 530 | ||
XM_006243021.2 | XP_006243083.1 | 5 | 544 | |||
GenBank mRNA | CB583270.1 | - | 5 | 530 | ||
D49395.1 | - | 5 | 421 |
