siRNA Details
Gene Symbol: Actn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031005.3 | NP_112267.1 | 9 | 992 | ||
GenBank mRNA | AF115386.1 | - | 9 | 992 | ||
AY437436.1 | - | 9 | 783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
