siRNA Details
Gene Symbol: Mapk14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256142.2 | XP_006256204.1 | 9 | 722 | ||
GenBank mRNA | U73142.1 | - | 9 | 718 | ||
U91847.1 | - | 9 | 707 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001168514, NM_001168513, NM_011951, U10871, AF128892, and others.
Gene Ontology Categories:
