siRNA Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031151.2 | NP_112413.2 | 2 | 307 | ||
GenBank mRNA | BC063165.1 | - | 2 | 307 | 6918633 | |
DQ402951.1 | - | 1 | 141 | |||
FQ213494.1 | - | 2 | 260 | |||
FQ213817.1 | - | 2 | 224 | |||
FQ215385.1 | - | 2 | 267 | |||
FQ215647.1 | - | 2 | 268 | |||
FQ216782.1 | - | 2 | 298 | |||
FQ217232.1 | - | 2 | 259 | |||
FQ224847.1 | - | 2 | 204 | |||
FQ224979.1 | - | 2 | 286 | |||
X04240.1 | - | 2 | 248 |
