siRNA Details
Gene Symbol: Mdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031151.2 | NP_112413.2 | 4 | 514 | ||
GenBank mRNA | BC063165.1 | - | 4 | 514 | 6918633 | |
DQ402951.1 | - | 3 | 348 | |||
FQ213494.1 | - | 4 | 467 | |||
FQ213817.1 | - | 4 | 429 | |||
FQ215385.1 | - | 4 | 474 | |||
FQ215647.1 | - | 4 | 475 | |||
FQ216782.1 | - | 4 | 505 | |||
FQ217232.1 | - | 4 | 466 | |||
FQ224847.1 | - | 4 | 411 | |||
FQ224979.1 | - | 4 | 493 | |||
X04240.1 | - | 4 | 455 |
