siRNA Details
Gene Symbol: LOC103689931 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006246345.2 | XP_006246407.1 | 8 | 1757 | ||
XM_008767732.1 | XP_008765954.1 | 7 | 1616 |
Gene Symbol: Hnrnpab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031330.2 | NP_112620.2 | 7 | 1562 | ||
XM_003750817.3 | XP_003750865.1 | 8 | 1757 | |||
GenBank mRNA | BC066664.1 | - | 7 | 1557 | 5621982 | |
CK473681.1 | - | 1 | 543 | |||
FQ221972.1 | - | 1 | 260 |
Gene Symbol: LOC100910156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ212801.1 | - | 1 | 420 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001048061, NM_010448, BC043069, AK076132, CF751043, and others.
Gene Ontology Categories:
