siRNA Details
Gene Symbol: RNF24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134337.2 | NP_001127809.1 | 6 | 1184 | ||
NM_001134338.2 | NP_001127810.1 | 7 | 1253 | |||
NM_001321749.1 | NP_001308678.1 | 6 | 1420 | |||
NM_007219.4 | NP_009150.1 | 6 | 1476 | |||
XM_011529145.2 | XP_011527447.1 | 8 | 1499 | |||
XM_011529146.2 | XP_011527448.1 | 7 | 1429 | |||
XM_017027620.1 | XP_016883109.1 | 7 | 1521 | |||
XM_017027621.1 | XP_016883110.1 | 7 | 1290 | |||
XM_017027622.1 | XP_016883111.1 | 8 | 1583 | |||
GenBank mRNA | AF151081.1 | - | 1 | 571 | ||
AL079313.1 | - | 4 | 892 | |||
AL096778.1 | - | 6 | 1274 | |||
BC039584.1 | - | 6 | 1103 | 5214313 |
