siRNA Details
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031665.2 | NP_113853.1 | 7 | 829 | ||
GenBank mRNA | BC081769.1 | - | 7 | 829 | 7134641 | |
U56815.1 | - | 7 | 805 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021433, AK019106, AK044905, AK031673, AK018167, and others.
Gene Ontology Categories:
