siRNA Details
Gene Symbol: Tagln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035236.1 | NP_001030313.1 | 3 | 427 | ||
NM_031676.2 | NP_113864.2 | 2 | 375 | |||
GenBank mRNA | AF459788.1 | - | 3 | 571 | ||
FQ212703.1 | - | 2 | 369 | |||
FQ212858.1 | - | 2 | 369 | |||
FQ214377.1 | - | 2 | 368 | |||
FQ214483.1 | - | 2 | 369 | |||
FQ214564.1 | - | 2 | 369 | |||
M84725.1 | - | 2 | 349 |
Gene Symbol: LOC103693564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768676.1 | XP_008766898.1 | 3 | 644 |

More Information
Other Species Targeted

Gene Ontology Categories:
