siRNA Details
Gene Symbol: B4GALT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007255.2 | NP_009186.1 | 6 | 1518 | ||
XM_006714816.3 | XP_006714879.1 | 6 | 1450 | |||
XM_017008999.1 | XP_016864488.1 | 6 | 3641 | |||
GenBank mRNA | AB028600.1 | - | 6 | 1465 | ||
AF142675.1 | - | 6 | 1458 | |||
AK022566.1 | - | 4 | 2525 | |||
AK023506.1 | - | 6 | 1461 | |||
AK095882.1 | - | 6 | 2122 | |||
AK123092.1 | - | 2 | 2244 | |||
BC007317.2 | - | 6 | 1518 | 3049998 | ||
BC062983.1 | - | 6 | 1485 | 4577572 | ||
BC072403.1 | - | 6 | 1507 | 6094074 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
