siRNA Details
Gene Symbol: Htra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031721.1 | NP_113909.1 | 3 | 754 | ||
GenBank mRNA | AF179370.1 | - | 3 | 754 | ||
BC081767.1 | - | 3 | 779 | 7121959 | ||
FQ228889.1 | - | 2 | 206 | |||
FQ228942.1 | - | 2 | 232 |
