siRNA Details
Gene Symbol: Abcb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031760.1 | NP_113948.1 | 6 | 498 | ||
GenBank mRNA | AF010597.1 | - | 6 | 471 | ||
U69487.1 | - | 6 | 498 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Abcb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031760.1 | NP_113948.1 | 6 | 498 | ||
GenBank mRNA | AF010597.1 | - | 6 | 471 | ||
U69487.1 | - | 6 | 498 |