siRNA Details
Gene Symbol: Ybx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031979.3 | NP_114185.3 | 5 | 627 | ||
GenBank mRNA | D28557.1 | - | 5 | 491 | ||
U22893.1 | - | 5 | 630 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
