siRNA Details
Gene Symbol: SYNPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007286.5 | NP_009217.3 | 3 | 4754 | ||
XM_005268370.1 | XP_005268427.1 | 3 | 4697 | |||
XM_005268371.1 | XP_005268428.1 | 3 | 4617 | |||
XM_006714755.3 | XP_006714818.1 | 4 | 5306 | |||
XM_017009007.1 | XP_016864496.1 | 4 | 5339 | |||
XM_017009008.1 | XP_016864497.1 | 4 | 5278 | |||
GenBank mRNA | AB028952.1 | - | 3 | 4712 | ||
AL049959.1 | - | 1 | 1136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
