siRNA Details
Gene Symbol: Dnaja2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032079.2 | NP_114468.2 | 1, 2 | 94 | ||
GenBank mRNA | BC087010.1 | - | 1, 2 | 94 | 7191358 | |
U95727.1 | - | 1, 2 | 128 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019794, AK083208, AB028853, BC003420, AK169169, and others.
Gene Ontology Categories:
