siRNA Details
Gene Symbol: Ogfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053340.2 | NP_445792.2 | 7 | 2033 | ||
GenBank mRNA | BC101898.1 | - | 7 | 2014 | 7932932 | |
FQ230610.1 | - | 1 | 890 | |||
FQ232690.1 | - | 1 | 890 |

More Information
Other Species Targeted

Gene Ontology Categories:
