siRNA Details
Gene Symbol: Ngfrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053401.1 | NP_445853.1 | 2 | 135 | ||
XM_006257257.2 | XP_006257319.1 | 3 | 335 | |||
XM_006257258.2 | XP_006257320.1 | 3 | 249 | |||
GenBank mRNA | AF187065.1 | - | 2 | 135 | ||
AY833556.1 | - | 3 | 240 | |||
BC058503.1 | - | 3 | 339 | 6887969 | ||
BC166418.1 | - | 3 | 248 | 7314222 |

More Information
Other Species Targeted

Gene Ontology Categories:
