siRNA Details
Gene Symbol: Ngfrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053401.1 | NP_445853.1 | 2 | 141 | ||
XM_006257257.2 | XP_006257319.1 | 3 | 341 | |||
XM_006257258.2 | XP_006257320.1 | 3 | 255 | |||
GenBank mRNA | AF187065.1 | - | 2 | 141 | ||
AY833556.1 | - | 3 | 246 | |||
BC058503.1 | - | 3 | 345 | 6887969 | ||
BC166418.1 | - | 3 | 254 | 7314222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
