siRNA Details
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053424.1 | NP_445876.1 | 26 | 3444 | ||
XM_006250788.2 | XP_006250850.1 | 25 | 3807 | |||
XM_006250789.2 | XP_006250851.1 | 26 | 3904 | |||
XM_006250790.2 | XP_006250852.1 | 26 | 3525 | |||
XM_006250791.2 | XP_006250853.1 | 25 | 3359 | |||
GenBank mRNA | AF004017.1 | - | 23 | 3282 | ||
AF027362.1 | - | 23 | 3314 | |||
AF124441.1 | - | 25 | 3347 | |||
AF254802.1 | - | 26 | 3444 |
