siRNA Details
Gene Symbol: PLA2R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195641.1 | NP_001182570.1 | 30 | 5071 | ||
NM_007366.4 | NP_031392.3 | 30 | 5077 | |||
XM_017003600.1 | XP_016859089.1 | 20 | 3188 | |||
XM_017003602.1 | XP_016859091.1 | 19 | 2914 | |||
GenBank mRNA | U17033.1 | - | 30 | 5077 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
