siRNA Details
Gene Symbol: Slc22a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053537.2 | NP_445989.2 | 2 | 432 | ||
GenBank mRNA | AY816233.1 | - | 2 | 432 | ||
L27651.1 | - | 2 | 492 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc22a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053537.2 | NP_445989.2 | 2 | 432 | ||
GenBank mRNA | AY816233.1 | - | 2 | 432 | ||
L27651.1 | - | 2 | 492 |