siRNA Details
Gene Symbol: SHOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001269039.2 | NP_001255968.1 | 8 | 2766 | ||
NM_001324336.1 | NP_001311265.1 | 9 | 2861 | |||
NM_001324337.1 | NP_001311266.1 | 10 | 3089 | |||
NM_007373.3 | NP_031399.2 | 9 | 2904 | |||
NR_136749.1 | 8 | 1967 | ||||
XM_017016702.1 | XP_016872191.1 | 9 | 2809 | |||
XM_017016703.1 | XP_016872192.1 | 9 | 8626 | |||
XM_017016704.1 | XP_016872193.1 | 9 | 2808 | |||
GenBank mRNA | AB020669.1 | - | 9 | 2832 | ||
AK090820.1 | - | 7 | 1923 | |||
BC044752.1 | - | 9 | 2840 | |||
BC050445.1 | - | 8 | 2759 | 6067669 | ||
BX649055.1 | - | 10 | 2671 |
