siRNA Details
Gene Symbol: Madd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053585.1 | NP_446037.1 | 3 | 435 | ||
GenBank mRNA | U72995.1 | - | 3 | 435 |

More Information
Other Species Targeted

Gene Ontology Categories:
