siRNA Details
Gene Symbol: Fez2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053600.2 | NP_446052.2 | 8 | 1114 | ||
XM_006239637.2 | XP_006239699.1 | 7 | 1174 | |||
XM_006239638.2 | XP_006239700.1 | 7 | 1084 | |||
GenBank mRNA | AB076183.1 | - | 8 | 1114 | ||
AB080086.1 | - | 7 | 1012 | |||
AF120109.1 | - | 3 | 207 | |||
AF120110.1 | - | 2 | 126 | |||
AF120111.1 | - | 2 | 126 | |||
BC100060.1 | - | 7 | 1014 | 7128839 | ||
CB725415.1 | - | 4 | 356 | |||
U64689.1 | - | 8 | 940 |
