siRNA Details
Gene Symbol: Usp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053774.2 | NP_446226.2 | 2 | 459 | ||
XM_006242861.2 | XP_006242923.1 | 2 | 620 | |||
GenBank mRNA | AF106659.1 | - | 2 | 459 | ||
AF202453.1 | - | 1 | 332 | |||
BC085719.1 | - | 2 | 433 | 7193298 |
