siRNA Details
Gene Symbol: PRDX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012094.4 | NP_036226.1 | 2 | 394 | ||
NM_181651.2 | NP_857634.1 | 2 | 394 | |||
GenBank mRNA | AF110731.1 | - | 2 | 302 | ||
AF112212.1 | - | 2 | 293 | |||
AF124993.1 | - | 2 | 335 | |||
AF197952.1 | - | 2 | 375 | |||
AF231705.1 | - | 2 | 285 | |||
AF242525.1 | - | 2 | 301 | |||
AJ249483.1 | - | 1 | 110 | |||
BC110983.1 | - | 2 | 394 | 5166149 | ||
BC113723.1 | - | 2 | 300 | 8322775 | ||
BC113725.1 | - | 2 | 300 | 8322777 | ||
BC143849.1 | - | 2 | 300 | 9052365 | ||
BU598032.1 | - | 2 | 388 | |||
CR457203.1 | - | 2 | 266 | |||
FJ224298.1 | - | 1 | 110 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
