siRNA Details
Gene Symbol: Pcdha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053941.1 | NP_446393.1 | 1 | 915 | ||
GenBank mRNA | AB113386.1 | - | 1 | 915 | ||
AF539749.1 | - | 1 | 538 | |||
AY573977.1 | - | 1 | 915 |
