siRNA Details
Gene Symbol: Slc4a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270860.1 | NP_001257789.1 | 8 | 1387 | ||
NM_001270861.1 | NP_001257790.1 | 8 | 1426 | |||
NM_058211.2 | NP_478118.1 | 8 | 1426 | |||
XM_008770481.1 | XP_008768703.1 | 7 | 1073 | |||
XM_008770482.1 | XP_008768704.1 | 7 | 1034 | |||
XM_008770483.1 | XP_008768705.1 | 7 | 1073 | |||
GenBank mRNA | AF069511.1 | - | 8 | 1387 | ||
AF070475.1 | - | 8 | 1426 | |||
AF080106.1 | - | 8 | 1426 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001033270, HM624051, AK153567, JQ349032, JQ344322, and others.
Gene Ontology Categories:
