siRNA Details
Gene Symbol: Nr2f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080778.2 | NP_542956.1 | 3 | 1527 | ||
XM_006229366.2 | XP_006229428.1 | 3 | 6414 | |||
GenBank mRNA | AF003944.1 | - | 3 | 1528 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_183261, NM_009697, BC094360, AK032882, BC042484, and others.
Gene Ontology Categories:
