siRNA Details
Gene Symbol: LOC291863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013889.1 | NP_001013911.1 | 2 | 104 | ||
GenBank mRNA | BC078681.1 | - | 2 | 104 | 7123712 |
Gene Symbol: Ces1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133295.3 | NP_579829.3 | 2 | 129 | ||
GenBank mRNA | AF171640.1 | - | 2 | 78 | ||
BC061789.1 | - | 2 | 129 | 5623732 | ||
L46791.1 | - | 2 | 108 | |||
L81144.1 | - | 2 | 88 | |||
X51974.1 | - | 2 | 126 |

More Information
Other Species Targeted

Gene Ontology Categories:
