siRNA Details
Gene Symbol: Ces1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133295.3 | NP_579829.3 | 14 | 1903 | ||
GenBank mRNA | BC061789.1 | - | 14 | 1903 | 5623732 | |
CF111142.1 | - | 11 | 1403 | |||
L46791.1 | - | 14 | 1883 | |||
X51974.1 | - | 14 | 1900 |
Gene Symbol: LOC291863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013889.1 | NP_001013911.1 | 15 | 1878 | ||
GenBank mRNA | BC078681.1 | - | 15 | 1878 | 7123712 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
