siRNA Details
Gene Symbol: DNAI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281428.1 | NP_001268357.1 | 2, 3 | 329 | ||
NM_012144.3 | NP_036276.1 | 2, 3 | 329 | |||
XM_006716758.3 | XP_006716821.1 | 1, 2 | 27 | |||
XM_011517846.1 | XP_011516148.1 | 2, 3 | 329 | |||
XM_011517847.2 | XP_011516149.1 | 2, 3 | 329 | |||
XM_011517848.1 | XP_011516150.1 | 2, 3 | 329 | |||
XM_011517849.1 | XP_011516151.1 | 2, 3 | 329 | |||
XM_011517850.2 | XP_011516152.1 | 2, 3 | 329 | |||
XM_017014625.1 | XP_016870114.1 | 2, 3 | 329 | |||
GenBank mRNA | AF091619.1 | - | 2, 3 | 246 | ||
AK290696.1 | - | 2, 3 | 253 | |||
AK302499.1 | - | 2, 3 | 192 | |||
AK303937.1 | - | 2, 3 | 218 | |||
AK314639.1 | - | 2, 3 | 241 | |||
BC030583.2 | - | 2, 3 | 257 | 4837991 | ||
BM013886.1 | - | 2, 3 | 268 | |||
DB033871.1 | - | 2, 3 | 253 | |||
DB046118.1 | - | 2, 3 | 330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
