siRNA Details
Gene Symbol: F8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012151.3 | NP_036283.2 | 1 | 1019 | ||
GenBank mRNA | BC039693.1 | - | 1 | 1005 | 5214628 | |
BC071963.1 | - | 1 | 1009 | 5477018 |
Gene Symbol: F8A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007523.1 | NP_001007524.1 | 1 | 1004 |
Gene Symbol: F8A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007524.1 | NP_001007525.1 | 1 | 1004 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
