siRNA Details
Gene Symbol: Rab3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133536.2 | NP_598220.1 | 4, 5 | 582 | ||
XM_008760719.1 | XP_008758941.1 | 4, 5 | 747 | |||
GenBank mRNA | D78197.1 | - | 4, 5 | 559 | ||
U54807.1 | - | 4, 5 | 586 |
