siRNA Details
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134349.3 | NP_599176.1 | 4 | 645 | ||
GenBank mRNA | BC063150.1 | - | 4 | 643 | 6922267 | |
FQ209427.1 | - | 4 | 666 | |||
FQ209464.1 | - | 4 | 658 | |||
FQ209465.1 | - | 4 | 665 | |||
FQ211049.1 | - | 4 | 665 | |||
FQ212841.1 | - | 4 | 666 | |||
FQ219410.1 | - | 4 | 639 | |||
FQ219464.1 | - | 4 | 671 | |||
FQ219581.1 | - | 4 | 667 | |||
FQ219850.1 | - | 4 | 657 | |||
J03752.1 | - | 4 | 629 |
