siRNA Details
Gene Symbol: FBXW4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033408.1 | 1 | 740 | |||
GenBank mRNA | AF174606.1 | - | 1 | 740 | ||
AK024483.1 | - | 1 | 3053 | |||
AK314175.1 | - | 1 | 603 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
