siRNA Details
Gene Symbol: Ptprg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134356.1 | NP_599183.1 | 30 | 4415 | ||
GenBank mRNA | AY177703.1 | - | 30 | 4415 | ||
AY177704.1 | - | 29 | 4328 |

More Information
Other Species Targeted

Gene Ontology Categories:
