siRNA Details
Gene Symbol: Uxs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139336.1 | NP_647552.1 | 14 | 1141 | ||
XM_006244296.2 | XP_006244358.1 | 14 | 1199 | |||
XM_008766751.1 | XP_008764973.1 | 15 | 1657 | |||
GenBank mRNA | AF482705.1 | - | 14 | 1141 | ||
BC086988.1 | - | 14 | 1152 | 7192831 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026430, BC037049, AK005536, AK152376, AK184776, and others.
Gene Ontology Categories:
