siRNA Details
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153730.2 | NP_714952.1 | 1 | 122 | ||
XM_006249843.2 | XP_006249905.1 | 2 | 250 | |||
XM_006249844.2 | XP_006249906.1 | 2 | 524 | |||
XM_006249845.1 | XP_006249907.1 | 2 | 251 | |||
XM_006249846.2 | XP_006249908.1 | 2 | 249 | |||
XM_008769539.1 | XP_008767761.1 | 2 | 289 | |||
GenBank mRNA | AJ133126.1 | - | 1 | 164 | ||
BC086983.1 | - | 2 | 252 | 7133597 | ||
FQ210179.1 | - | 2 | 233 |

More Information
Other Species Targeted

Gene Ontology Categories:
