siRNA Details
Gene Symbol: Hsbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173119.2 | NP_775142.1 | 4 | 423 | ||
GenBank mRNA | AF522937.1 | - | 4 | 370 | ||
BC058131.1 | - | 4 | 408 | 6919748 | ||
EX491168.1 | - | 4 | 423 | |||
FQ211891.1 | - | 4 | 391 | |||
FQ212621.1 | - | 4 | 395 | |||
FQ213728.1 | - | 4 | 365 | |||
FQ216175.1 | - | 4 | 402 | |||
FQ229155.1 | - | 4 | 408 | |||
FQ229754.1 | - | 4 | 382 | |||
FQ230047.1 | - | 4 | 380 | |||
FQ230722.1 | - | 4 | 382 | |||
FQ233773.1 | - | 4 | 381 |
