siRNA Details
Gene Symbol: Ncstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174864.3 | NP_777353.1 | 2 | 210 | ||
XM_006250283.1 | XP_006250345.1 | 2 | 189 | |||
XM_008769733.1 | XP_008767955.1 | 2 | 195 | |||
XM_008769734.1 | XP_008767956.1 | 1 | 37 | |||
XM_008769735.1 | XP_008767957.1 | 1 | 37 | |||
GenBank mRNA | AF510722.1 | - | 2 | 118 | ||
AY101378.1 | - | 2 | 118 | |||
BC074006.1 | - | 2 | 210 | |||
BC167747.1 | - | 2 | 189 | 8365089 |
