siRNA Details
Gene Symbol: Ncstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174864.3 | NP_777353.1 | 3, 4 | 400 | ||
XM_008769733.1 | XP_008767955.1 | 3, 4 | 385 | |||
XM_008769734.1 | XP_008767956.1 | 3, 4 | 342 | |||
GenBank mRNA | AY101378.1 | - | 3, 4 | 308 | ||
BC074006.1 | - | 4, 5 | 515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
