siRNA Details
Gene Symbol: Taar4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175583.1 | NP_783173.1 | 1 | 745 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
