siRNA Details
Gene Symbol: Cacna2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175592.2 | NP_783182.1 | 23, 24 | 2011 | ||
XM_006243734.2 | XP_006243796.1 | 23, 24 | 2285 | |||
XM_006243735.2 | XP_006243797.1 | 23, 24 | 2285 | |||
GenBank mRNA | AF486277.1 | - | 23, 24 | 2011 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_020263, NM_001174047, AK164143, AF247141, BC056389, and others.
Gene Ontology Categories:
