siRNA Details
Gene Symbol: Slc4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178092.2 | NP_835193.1 | 8, 9 | 1011 | ||
XM_006234275.2 | XP_006234337.1 | 8, 9 | 1562 | |||
XM_006234276.2 | XP_006234338.1 | 8, 9 | 1559 | |||
XM_006234277.2 | XP_006234339.1 | 8, 9 | 1342 | |||
XM_006234279.2 | XP_006234341.1 | 8, 9 | 1557 | |||
GenBank mRNA | AF439856.1 | - | 8, 9 | 1011 | ||
AY579373.1 | - | 8, 9 | 1014 | |||
JX073716.2 | - | 8, 9 | 1055 | |||
JX073718.2 | - | 8, 9 | 1055 | |||
KF736947.1 | - | 8, 9 | 946 | |||
KF736949.1 | - | 8, 9 | 979 |
