siRNA Details
Gene Symbol: Tsg101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181628.2 | NP_853659.2 | 8 | 824 | ||
XM_008759341.1 | XP_008757563.1 | 6 | 725 | |||
GenBank mRNA | AY293306.1 | - | 8 | 783 | ||
BC070951.1 | - | 8 | 824 | 7100763 | ||
FQ231102.1 | - | 5 | 504 |
