siRNA Details
Gene Symbol: Taf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037310.1 | NP_001032387.1 | 3 | 1097 | ||
NM_184048.2 | NP_908937.2 | 3 | 618 | |||
GenBank mRNA | AY325235.1 | - | 2 | 454 | ||
AY359819.1 | - | 1 | 414 | |||
BC091109.1 | - | 3 | 607 | 7322630 | ||
FQ213120.1 | - | 3 | 601 | |||
FQ215462.1 | - | 3 | 612 | |||
FQ229736.1 | - | 3 | 611 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
